Pyrrole-3-carboxylic acid hydrate, 95%
Pyrrole-3-carboxylic acid hydrate, 95%
Pyrrole-3-carboxylic acid hydrate, 95%
Pyrrole-3-carboxylic acid hydrate, 95%
Thermo Scientific Chemicals
Pyrrole-3-carboxylic acid hydrate, 95%
CAS: 336100-46-0 | C5H5NO2 | 111.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
Packaging:
Glass bottle
Catalog number FSA377655000
Price (MYR)
796.00
MG
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
796.00
MG
Add to cart
Specifications
Chemical Name or MaterialPyrrole-3-carboxylic acid hydrate
CAS336100-46-0
Health Hazard 1GHS Signal Word: Warning
Melting Point144.0°C to 147.0°C
ColorBeige to Brown