2-Naphthoyl chloride, 98%
2-Naphthoyl chloride, 98%
2-Naphthoyl chloride, 98%
2-Naphthoyl chloride, 98%
Thermo Scientific Chemicals
2-Naphthoyl chloride, 98%
CAS: 2243-83-6 | C11H7ClO | 190.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA376590500
Price (MYR)
1,951.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,951.00
EA
Add to cart
Specifications
Boiling Point156.0°C to 158.0°C (7.0mmHg)
Chemical Name or Material2-Naphthoyl chloride
Melting Point49.0°C to 54.0°C
CAS2243-83-6
ColorBrown to Gray