p-Toluidine, 99%, crystalline molten mass
p-Toluidine, 99%, crystalline molten mass
p-Toluidine, 99%, crystalline molten mass
p-Toluidine, 99%, crystalline molten mass
Thermo Scientific Chemicals
p-Toluidine, 99%, crystalline molten mass
CAS: 106-49-0 | C7H9N | 107.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number FSA376352500
Price (MYR)
249.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
249.00
EA
Add to cart
Specifications
Chemical Name or Materialp-Toluidine
Name Note99%
CAS106-49-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic in contact with skin.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
Toxic if inhaled.
Very toxic to aquatic life.