3,5-Dichloro-1,2,4-thiadiazole, 97%
3,5-Dichloro-1,2,4-thiadiazole, 97%
3,5-Dichloro-1,2,4-thiadiazole, 97%
3,5-Dichloro-1,2,4-thiadiazole, 97%
Thermo Scientific Chemicals
3,5-Dichloro-1,2,4-thiadiazole, 97%
CAS: 2254-88-8 | C2Cl2N2S | 155.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA376030250
Price (MYR)
4,339.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,339.00
EA
Add to cart
Specifications
Chemical Name or Material3,5-Dichloro-1,2,4-thiadiazole
CAS2254-88-8
Health Hazard 1GHS Signal Word: Warning
ColorColorless to Yellow
Boiling Point165.0°C to 167.0°C