Dicyclohexyl phthalate, 99%
Dicyclohexyl phthalate, 99%
Dicyclohexyl phthalate, 99%
Dicyclohexyl phthalate, 99%
Thermo Scientific Chemicals
Dicyclohexyl phthalate, 99%
CAS: 84-61-7 | C20H26O4 | 330.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number FSA375052500
Price (MYR)
451.00
G
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
451.00
G
Add to cart
Specifications
Chemical Name or MaterialDicyclohexyl phthalate
CAS84-61-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
May damage the unborn child.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention