Mono-methyl phthalate, 98%
Mono-methyl phthalate, 98%
Mono-methyl phthalate, 98%
Mono-methyl phthalate, 98%
Thermo Scientific Chemicals
Mono-methyl phthalate, 98%
CAS: 4376-18-5 | C9H8O4 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA374880250
Price (MYR)
1,530.00
G
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,530.00
G
Add to cart
Specifications
Chemical Name or Materialmono-Methyl phthalate
CAS4376-18-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0