2-Iodoxybenzoic acid, stabilized
2-Iodoxybenzoic acid, stabilized
2-Iodoxybenzoic acid, stabilized
2-Iodoxybenzoic acid, stabilized
Thermo Scientific Chemicals
2-Iodoxybenzoic acid, stabilized
CAS: 61717-82-6 | C7H5IO4 | 280.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA374650500
Price (MYR)
3,484.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,484.00
EA
Add to cart
Specifications
Chemical Name or Material2-Iodoxybenzoic acid
CAS61717-82-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
CAS64297-64-9
View more
oxidation reagent
RUO – Research Use Only