Potassium iodide, 99+%, for analysis
Potassium iodide, 99+%, for analysis
Potassium iodide, 99+%, for analysis
Potassium iodide, 99+%, for analysis
Thermo Scientific Chemicals
Potassium iodide, 99+%, for analysis
CAS: 7681-11-0 | IK | 166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA373655000
Price (MYR)
1,164.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,164.00
BOT
Add to cart
Specifications
Boiling Point1330.0°C
Chemical Name or MaterialPotassium iodide
Melting Point680.0°C
CAS7681-11-0
ColorTan to White