1-Methylpyrazole, 98%
1-Methylpyrazole, 98%
1-Methylpyrazole, 98%
1-Methylpyrazole, 98%
Thermo Scientific Chemicals
1-Methylpyrazole, 98%
CAS: 930-36-9 | C4H6N2 | 82.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA373540500
Price (MYR)
1,621.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,621.00
EA
Add to cart
Specifications
Chemical Name or Material1-Methylpyrazole
CAS930-36-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0