Bromodiphenylmethane, 90%
Bromodiphenylmethane, 90%
Bromodiphenylmethane, 90%
Bromodiphenylmethane, 90%
Thermo Scientific Chemicals
Bromodiphenylmethane, 90%
CAS: 776-74-9 | C13H11Br | 247.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA373465000
Price (MYR)
1,145.00
G
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,145.00
G
Add to cart
Specifications
Chemical Name or MaterialBromodiphenylmethane
CAS776-74-9
Health Hazard 1GHS Signal Word: Danger
Melting Point33.0°C to 42.0°C
EINECS Number212-279-8