4-Mercaptobenzoic acid, 90%
4-Mercaptobenzoic acid, 90%
4-Mercaptobenzoic acid, 90%
4-Mercaptobenzoic acid, 90%
Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%
CAS: 1074-36-8 | C7H6O2S | 154.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA373300250
Price (MYR)
2,371.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,371.00
EA
Add to cart
Specifications
Chemical Name or Material4-Mercaptobenzoic acid
CAS1074-36-8
CAS Min %88.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0