4,4'-Dimethyl-2,2'-bipyridyl, 99+%
4,4'-Dimethyl-2,2'-bipyridyl, 99+%
4,4'-Dimethyl-2,2'-bipyridyl, 99+%
4,4'-Dimethyl-2,2'-bipyridyl, 99+%
Thermo Scientific Chemicals
4,4'-Dimethyl-2,2'-bipyridyl, 99+%
CAS: 1134-35-6 | C12H12N2 | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA373230250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4, 4'-Dimethyl-2, 2'-bipyridyl
CAS1134-35-6
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0