2-Cyanobenzaldehyde, 98%
2-Cyanobenzaldehyde, 98%
2-Cyanobenzaldehyde, 98%
2-Cyanobenzaldehyde, 98%
Thermo Scientific Chemicals
2-Cyanobenzaldehyde, 98%
CAS: 7468-67-9 | C8H5NO | 131.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA371860050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Cyanobenzaldehyde
CAS7468-67-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0