cis-2,6-Dimethylmorpholine, 97%
cis-2,6-Dimethylmorpholine, 97%
cis-2,6-Dimethylmorpholine, 97%
cis-2,6-Dimethylmorpholine, 97%
Thermo Scientific Chemicals
cis-2,6-Dimethylmorpholine, 97%
CAS: 6485-55-8 | C6H13NO | 115.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA371840250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialcis-2, 6-Dimethylmorpholine
CAS6485-55-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa