Hydroxylapatite, for analysis
Hydroxylapatite, for analysis
Hydroxylapatite, for analysis
Hydroxylapatite, for analysis
Thermo Scientific Chemicals
Hydroxylapatite, for analysis
CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA371261000
Price (MYR)
486.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
486.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydroxylapatite
Melting Point1700.0°C
CAS1306-06-5
ColorWhite
Physical FormPowder