1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
1,8-Diaminonaphthalene, 97%
Thermo Scientific Chemicals
1,8-Diaminonaphthalene, 97%
CAS: 479-27-6 | C10H10N2 | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA371181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 8-Diaminonaphthalene
Name Note97%
CAS479-27-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning