1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
Thermo Scientific Chemicals
1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
CAS: 54149-17-6 | C5H11BrO2 | 183.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA370980100
Price (MYR)
599.00
G
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
599.00
G
Add to cart
Specifications
Chemical Name or Material1-Bromo-2-(2-methoxyethoxy)ethane
Name Note90%
CAS54149-17-6
CAS Min %88.0
CAS Max %100.0