Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Sand, pure, 40-100 mesh
Thermo Scientific Chemicals
Sand, pure, 40-100 mesh
CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number FSA370942500
Price (MYR)
260.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
260.00
EA
Add to cart
Specifications
Boiling Point2230.0°C
Chemical Name or MaterialSand
Melting Point1610.0°C
Name Note40 to 100 Mesh
CAS14808-60-7
View more
Filter aid
RUO – Research Use Only