Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Thermo Scientific Chemicals
Ammonium bicarbonate, 98%, pure
CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA370930010
Price (MYR)
462.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
462.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium bicarbonate
Melting Point105°C
CAS1066-33-7
ColorWhite
Health Hazard 1GHS Signal Word: Warning