Phenylacetaldehyde, 98%, stabilized
Phenylacetaldehyde, 98%, stabilized
Phenylacetaldehyde, 98%, stabilized
Phenylacetaldehyde, 98%, stabilized
Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized
CAS: 122-78-1 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA370911000
Price (MYR)
348.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
348.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenylacetaldehyde
Name Note98%
Acidity5mg KOH/g max.
CAS122-78-1
Health Hazard 1GHS Signal Word: Warning