Iodine monochloride, 1M solution in dichloromethane
Iodine monochloride, 1M solution in dichloromethane
Iodine monochloride, 1M solution in dichloromethane
Iodine monochloride, 1M solution in dichloromethane
Thermo Scientific Chemicals
Iodine monochloride, 1M solution in dichloromethane
CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA370741000
Price (MYR)
471.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
471.00
BOT
Add to cart
Chemical Identifiers
CAS7790-99-0, 75-09-2
Specifications
Chemical Name or MaterialIodine monochloride
Name Note1, 0M solution in dichloromethane
CAS7790-99-0
ColorBrown to Orange
Health Hazard 1GHS Signal Word: Danger
View more
C-X bond formation reagent
RUO – Research Use Only