Sodium percarbonate, 13-14% active oxygen
Sodium percarbonate, 13-14% active oxygen
Sodium percarbonate, 13-14% active oxygen
Sodium percarbonate, 13-14% active oxygen
Thermo Scientific Chemicals
Sodium percarbonate, 13-14% active oxygen
CAS: 15630-89-4 | 1·5 H2O2 | 157.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA370730025
Price (MYR)
713.00
KG
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
713.00
KG
Add to cart
Specifications
Chemical Name or MaterialSodium percarbonate
CAS15630-89-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
View more
Oxidation reagent
RUO – Research Use Only