Sodium, dry stick
Sodium, dry stick
Sodium, dry stick
Thermo Scientific Chemicals
Sodium, dry stick
CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA370212500
Price (MYR)
958.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
958.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium
ColorGray or White
CAS7440-23-5
Melting Point98.0°C
Boiling Point883.0°C