2-(Bromomethyl)pyridine hydrobromide, 98%
2-(Bromomethyl)pyridine hydrobromide, 98%
2-(Bromomethyl)pyridine hydrobromide, 98%
2-(Bromomethyl)pyridine hydrobromide, 98%
Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%
CAS: 31106-82-8 | C6H6BrN·HBr | 252.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA370110250
Price (MYR)
1,292.00
G
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,292.00
G
Add to cart
Specifications
Chemical Name or Material2-(Bromomethyl)pyridine hydrobromide
CAS31106-82-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0