Zinc acetate, 99%, pure
Zinc acetate, 99%, pure
Zinc acetate, 99%, pure
Zinc acetate, 99%, pure
Thermo Scientific Chemicals
Zinc acetate, 99%, pure
CAS: 557-34-6 | C4H6O4Zn | 183.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA370081000
Price (MYR)
1,836.00
G
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,836.00
G
Add to cart
Specifications
Chemical Name or MaterialZinc acetate
Melting Point83°C to 86°C
CAS557-34-6
ColorGray-Green to Yellow
Health Hazard 1GHS Signal Word: Danger