Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™
CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol
Have Questions?
Change view
Quantity:
100 mL
Catalog number FSA370051000
Price (MYR)
416.00
BOT
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
416.00
BOT
Add to cart
Chemical Identifiers
CAS7646-85-7
Specifications
Chemical Name or MaterialZinc chloride
ColorBlue-Gray or Colorless to Yellow
Name Note1.0M solution in diethyl ether
CAS7646-85-7
Health Hazard 1Danger