Sodium selenate, 98%
Sodium selenate, 98%
Sodium selenate, 98%
Sodium selenate, 98%
Thermo Scientific Chemicals
Sodium selenate, 98%
CAS: 13410-01-0 | Na2O4Se | 188.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA369961000
Price (MYR)
955.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
955.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium selenate
CAS13410-01-0
ColorWhite
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.