Amberlite™ IRA-402, Cl-form, ion-exchange resin
Amberlite™ IRA-402, Cl-form, ion-exchange resin
Amberlite™ IRA-402, Cl-form, ion-exchange resin
Amberlite™ IRA-402, Cl-form, ion-exchange resin
Thermo Scientific Chemicals
Amberlite™ IRA-402, Cl-form, ion-exchange resin
CAS: 52439-77-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA369942500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ IRA-402
Physical FormBeads
CAS52439-77-7
Particle SizeParticle size analysis: uniformity coefficient: <1.60, harmonic mean size: 0.600 to 0.750mm,% <0.300mm: <1.0%,%>1.180mm: <5.0%
PackagingPlastic bottle
View more
Ion exchange resin
RUO – Research Use Only