Cerium(III) chloride, 99.9%, pure, anhydrous
Cerium(III) chloride, 99.9%, pure, anhydrous
Cerium(III) chloride, 99.9%, pure, anhydrous
Cerium(III) chloride, 99.9%, pure, anhydrous
Thermo Scientific Chemicals
Cerium(III) chloride, 99.9%, pure, anhydrous
CAS: 7790-86-5 | CeCl3 | 246.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA369750250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point1727.0°C
Chemical Name or MaterialCerium(III) chloride
Melting Point848.0°C
Name NoteAnhdrous
CAS7790-86-5