Strontium chloride, 99.99%, (trace metal basis), anhydrous
Strontium chloride, 99.99%, (trace metal basis), anhydrous
Strontium chloride, 99.99%, (trace metal basis), anhydrous
Strontium chloride, 99.99%, (trace metal basis), anhydrous
Thermo Scientific Chemicals
Strontium chloride, 99.99%, (trace metal basis), anhydrous
CAS: 10476-85-4 | Cl2Sr | 158.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA369740250
Price (MYR)
1,422.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,422.00
BOT
Add to cart
Specifications
Boiling Point1250.0°C
Chemical Name or MaterialStrontium chloride
Melting Point873.0°C
CAS10476-85-4
ColorWhite
View more
for ultra trace analysis
RUO – Research Use Only