6-Aminoquinoline, 98%
6-Aminoquinoline, 98%
6-Aminoquinoline, 98%
6-Aminoquinoline, 98%
Thermo Scientific Chemicals
6-Aminoquinoline, 98%
CAS: 580-15-4 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA369640050
Price (MYR)
630.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
630.00
EA
Add to cart
Specifications
Chemical Name or Material6-Aminoquinoline
CAS580-15-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.