Methyl thiosalicylate, 97%
Methyl thiosalicylate, 97%
Methyl thiosalicylate, 97%
Methyl thiosalicylate, 97%
Thermo Scientific Chemicals
Methyl thiosalicylate, 97%
CAS: 4892-02-8 | C8H8O2S | 168.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA369600050
Price (MYR)
601.00
G
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
601.00
G
Add to cart
Specifications
Chemical Name or MaterialMethyl thiosalicylate
Name Note97%
CAS4892-02-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: