1-Decanesulfonic acid, sodium salt, 96%
1-Decanesulfonic acid, sodium salt, 96%
1-Decanesulfonic acid, sodium salt, 96%
1-Decanesulfonic acid, sodium salt, 96%
Thermo Scientific Chemicals
1-Decanesulfonic acid, sodium salt, 96%
1-Decanesulfonic acid, sodium salt, 96%, Quantity: 25g, Packaging: Glass bottle, White to Cream, Melting Point: >300 deg.C, Molecular Weight: 244.32, Percent Purity: 96%, Assay Percent Range: 96%, Beilstein: 04, III, 27, CAS: 13419-61-9, CAS Max %: 100.0, CAS Min %: 95.0 | CAS: 13419-61-9 | C10H21NaO3S | 244.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA369451000
Price (MYR)
3,913.00
G
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,913.00
G
Add to cart
Specifications
Chemical Name or Material1-Decanesulfonic acid, sodium salt
Name Note96%
CAS13419-61-9
CAS Min %95.0
CAS Max %100.0