Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
Thermo Scientific Chemicals
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
CAS: 79-37-8 | C2Cl2O2 | 126.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA369181000
Price (MYR)
472.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
472.00
BOT
Add to cart
Chemical Identifiers
CAS79-37-8, 75-09-2
Specifications
Chemical Name or MaterialOxalyl chloride
Name Note2.0M solution in dichloromethane
CAS79-37-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Reacts violently with water.
Contact with water liberates toxic gas.
Corrosive to t
View more
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only