4-Methoxyphenylacetyl chloride, 98%
4-Methoxyphenylacetyl chloride, 98%
4-Methoxyphenylacetyl chloride, 98%
4-Methoxyphenylacetyl chloride, 98%
4-Methoxyphenylacetyl chloride, 98%
CAS: 4693-91-8 | C9H9ClO2 | 184.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
Packaging:
Glass bottle
Catalog number FSA368930020
Price (MYR)
-
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Methoxyphenylacetyl chloride
CAS4693-91-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0