2-Aminoimidazole hemisulfate, 98+%
2-Aminoimidazole hemisulfate, 98+%
2-Aminoimidazole hemisulfate, 98+%
2-Aminoimidazole hemisulfate, 98+%
Thermo Scientific Chemicals
2-Aminoimidazole hemisulfate, 98+%
CAS: 1450-93-7 | C6H12N6O4S | 264.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Packaging:
Glass bottle
Catalog number FSA368860100
Price (MYR)
1,809.00
G
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,809.00
G
Add to cart
Specifications
Chemical Name or Material2-Aminoimidazole hemisulfate
CAS1450-93-7
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0