(Triisopropylsilyl)acetylene, 97%
(Triisopropylsilyl)acetylene, 97%
(Triisopropylsilyl)acetylene, 97%
(Triisopropylsilyl)acetylene, 97%
Thermo Scientific Chemicals
(Triisopropylsilyl)acetylene, 97%
CAS: 89343-06-6 | C11H22Si | 182.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA368740050
Price (MYR)
566.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
566.00
EA
Add to cart
Specifications
Chemical Name or Material(Triisopropylsilyl)acetylene
Name Note97%
CAS89343-06-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning