(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
Thermo Scientific Chemicals
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
CAS: 98327-87-8 | C44H32P2 | 622.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA368640050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(±)-2, 2'-Bis(diphenylphosphino)-1, 1'-binaphthyl
CAS98327-87-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0