3-Iodobenzonitrile, 97%
3-Iodobenzonitrile, 97%
3-Iodobenzonitrile, 97%
3-Iodobenzonitrile, 97%
3-Iodobenzonitrile, 97%
CAS: 69113-59-3 | C7H4IN | 229.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA368600500
Price (MYR)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Iodobenzonitrile
Name Note97%
CAS69113-59-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning