1-Methyl-2-pyrrolidinone, 99.5%, for HPLC
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC
CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA368451000
Price (MYR)
531.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
531.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24.0°C
Absorbance0.01 max. at 340nm, 0.01 max. at 450nm, 0.1 max. at 320nm, 0.5 max. at 300nm, 1 max. at 285nm
CAS872-50-4
CAS Min %99.4
View more
Suitable for battery materials development.
For HPLC
RUO – Research Use Only