2-Iodobenzaldehyde, 98%
2-Iodobenzaldehyde, 98%
2-Iodobenzaldehyde, 98%
2-Iodobenzaldehyde, 98%
Thermo Scientific Chemicals
2-Iodobenzaldehyde, 98%
CAS: 26260-02-6 | C7H5IO | 232.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA368170010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Iodobenzaldehyde
CAS26260-02-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0