Stearyl stearate, 98%
Stearyl stearate, 98%
Stearyl stearate, 98%
Stearyl stearate, 98%
Thermo Scientific Chemicals
Stearyl stearate, 98%
CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA368110050
Price (MYR)
3,901.00
G
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,901.00
G
Add to cart
Specifications
Chemical Name or MaterialStearyl stearate
Name Note98%
CAS2778-96-3
ColorWhite
EINECS Number220-476-5