Methyl linolenate, 99%
Methyl linolenate, 99%
Methyl linolenate, 99%
Methyl linolenate, 99%
Thermo Scientific Chemicals
Methyl linolenate, 99%
CAS: 301-00-8 | C19H32O2 | 292.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA368100050
Price (MYR)
2,863.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,863.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl linolenate
Name Note99%
CAS301-00-8
ColorColorless
Density0.8950g/mL