N,N-Diisopropylethylamine, 99.5+%
N,N-Diisopropylethylamine, 99.5+%
N,N-Diisopropylethylamine, 99.5+%
N,N-Diisopropylethylamine, 99.5+%
Thermo Scientific Chemicals
N,N-Diisopropylethylamine, 99.5+%
CAS: 7087-68-5 | C8H19N | 129.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
50 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA367841000
Price (MYR)
375.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
375.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Diisopropylethylamine
Name Note99.5+%
CAS7087-68-5
CAS Min %99.5
Health Hazard 1GHS Signal Word: Danger