5-Fluoroisatin, 98%
5-Fluoroisatin, 98%
5-Fluoroisatin, 98%
Thermo Scientific Chemicals
5-Fluoroisatin, 98%
CAS: 443-69-6 | C8H4FNO2 | 165.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA367700250
Price (MYR)
2,769.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,769.00
EA
Add to cart
Specifications
Chemical Name or Material5-Fluoroisatin
Name Note98%
CAS443-69-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning