1-(Tetrahydro-2-furoyl)piperazine, 98%
1-(Tetrahydro-2-furoyl)piperazine, 98%
1-(Tetrahydro-2-furoyl)piperazine, 98%
1-(Tetrahydro-2-furoyl)piperazine, 98%
Thermo Scientific Chemicals
1-(Tetrahydro-2-furoyl)piperazine, 98%
CAS: 63074-07-7 | C9H16N2O2 | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA367160050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-(Tetrahydro-2-furoyl)piperazine
Name Note98%
CAS63074-07-7
CAS Min %97.5
ColorColorless to Yellow