4-Hydroxycarbazole, 98+%
4-Hydroxycarbazole, 98+%
4-Hydroxycarbazole, 98+%
4-Hydroxycarbazole, 98+%
Thermo Scientific Chemicals
4-Hydroxycarbazole, 98+%
CAS: 52602-39-8 | C12H9NO | 183.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA366960010
Price (MYR)
263.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
263.00
EA
Add to cart
Specifications
Chemical Name or Material4-Hydroxycarbazole
CAS52602-39-8
CAS Min %98.0
CAS Max %100.0
Health Hazard 2GHS H Statement: