Tri-tert-butylphosphonium tetrafluoroborate, 97+%
Tri-tert-butylphosphonium tetrafluoroborate, 97+%
Tri-tert-butylphosphonium tetrafluoroborate, 97+%
Tri-tert-butylphosphonium tetrafluoroborate, 97+%
Thermo Scientific Chemicals
Tri-tert-butylphosphonium tetrafluoroborate, 97+%
CAS: 131274-22-1 | C12H28BF4P | 290.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA366940050
Price (MYR)
1,562.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,562.00
EA
Add to cart
Specifications
Chemical Name or MaterialTri-tert-butylphosphonium tetrafluoroborate
CAS131274-22-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.