Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A
Thermo Scientific Chemicals
Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A
Aluminum oxide, Al2O3, CAS Number-1344-28-1, abrasit, abrarex, abramax, abramant, alundum, Aluminum oxide, alpha-alumina, fasertonerde, Aluminum oxide, aloxite, 2.5kg, 2980.0 deg.C, White, 3.97g/mL, 215-691-6, 300ppm max. | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA366680010
Price (MYR)
739.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
739.00
BOT
Add to cart
Specifications
Boiling Point2980.0°C
Chemical Name or MaterialAluminum oxide
Melting Point2050.0°C
Name NoteActivated, Neutral, Brockmann I
CAS1344-28-1
View more
for column chromatography
RUO – Research Use Only