Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Thermo Scientific Chemicals
Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA365750010
Price (MYR)
661.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
661.00
BOT
Add to cart
Specifications
Boiling Point2980.0°C
Chemical Name or MaterialAluminum oxide
Melting Point2030.0°C
CAS1344-28-1
ColorWhite
View more
for column chromatography
RUO – Research Use Only